Skip to Content
Merck
All Photos(1)

Documents

R1277

Nunc® CryoTube® racks

CryoTube rack holds 40 cryogenic tubes, 1 rack/pk

Synonym(s):

cryogenic vial rack, cyrogenic tube rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103000
NACRES:
NB.22

packaging

pack of 1 ea

manufacturer/tradename

Nunc

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Nunc® CryoTube® racks

Note: The Nunc CryoTube rack interlocks with CryoTube starfoot cryogenic vials.

  • Storage and handling of cryogenic vials/cryogenic tubes
  • One-handed use when handling cryogenic vials

Legal Information

CryoTube is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service