Skip to Content
Merck
All Photos(1)

Key Documents

R1275

Sigma-Aldrich

Plate roller

secures sealing film onto plate, pkg of 1 ea

Synonym(s):

film sealing accessory, film sealing device, microplate

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CLP 45,900

CLP 45,900


Estimated to ship onMay 26, 2025



Select a Size

Change View
1 EA
CLP 45,900

About This Item

UNSPSC Code:
41122410
NACRES:
NB.15

CLP 45,900


Estimated to ship onMay 26, 2025


sterility

non-sterile

packaging

pkg of 1 ea

manufacturer/tradename

Excel Scientific RL PLT-01

color

none

General description

Consists of a rubber coated dowel with a little give. The rubber allows better conformance of the film around the well area when sealing plates (Ensures sealing film covers all wells of a multiwell plate).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Muhammad Yogi Pratama et al.
Scientific reports, 10(1), 3128-3128 (2020-02-23)
Normalization procedures for the qRT-PCR analysis of miRNA in biological samples are recommended to reduce the variability caused by pre-analytical factors. Since there is no universal standardized normalization strategy for miRNA qRT-PCR studies, we conducted a throughout study to evaluate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service