Skip to Content
Merck
All Photos(1)

Documents

PZ0311

Sigma-Aldrich

PF-05089771

≥98% (HPLC)

Synonym(s):

4-[2-(3-Amino-1H-pyrazol-4-yl)-4-chlorophenoxy]-5-chloro-2-fluoro-N-4-thiazolyl-benzenesulfonamide 4-methylbenzenesulfonate (1:1), 4-[2-(5-Amino-1H-pyrazol-4-yl)-4-chloro-phenoxy]-5-chloro-2-fluoro-N-thiazol-4-yl-benzenesulfonamide p-toluene sulfonate salt, PF-05089771 p-toluene sulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12Cl2FN5O3S2 · C7H8O3S
CAS Number:
Molecular Weight:
672.56
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=CC(C2=C(N)NN=C2)=C(OC3=C(Cl)C=C(S(NC4=CSC=N4)(=O)=O)C(F)=C3)C=C1.CC5=CC=C(S(=O)(O)=O)C=C5

InChI

1S/C18H12Cl2FN5O3S2.C7H8O3S/c19-9-1-2-14(10(3-9)11-6-24-25-18(11)22)29-15-5-13(21)16(4-12(15)20)31(27,28)26-17-7-30-8-23-17;1-6-2-4-7(5-3-6)11(8,9)10/h1-8,26H,(H3,22,24,25);2-5H,1H3,(H,8,9,10)

InChI key

NVKBPDYKPNYMDR-UHFFFAOYSA-N

Biochem/physiol Actions

PF-05089771 is a selective and potent inhibitor of the human voltage-gated sodium ion channel Nav1.7 with an IC50 value of 11 nM. PF-05089771 targets the voltage-sensing domain, preferentially interacting with, and stabilizing, inactivated channel conformations by interaction with the voltage-sensor domain (VSD) of Domain 4. Nav1.7 is expressed in sensory neurons and is critical for pain processing. PF-05089771 is being investigated for the treatment of chronic neuropathic pain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aristos J Alexandrou et al.
PloS one, 11(4), e0152405-e0152405 (2016-04-07)
Human genetic studies show that the voltage gated sodium channel 1.7 (Nav1.7) is a key molecular determinant of pain sensation. However, defining the Nav1.7 contribution to nociceptive signalling has been hampered by a lack of selective inhibitors. Here we report
Sooyeon Jo et al.
Molecular pharmacology, 97(6), 377-383 (2020-03-21)
PF-05089771 is an aryl sulfonamide Nav1.7 channel blocker that binds to the inactivated state of Nav1.7 channels with high affinity but binds only weakly to channels in the resting state. Such aryl sulfonamide Nav1.7 channel blockers bind to the extracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service