Skip to Content
Merck
All Photos(1)

Key Documents

PZ0184

Sigma-Aldrich

PF-04457845

≥98% (HPLC)

Synonym(s):

N-(3-pyridazinyl)-4-(3-[5-trifluoromethyl-2-pyridinyloxy]benzylidene)piperidine-1-carboxamide, N-3-Pyridazinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methylene]-1-piperidinecarboxamide, PF-4457845

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20F3N5O2
CAS Number:
Molecular Weight:
455.43
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL

storage temp.

room temp

SMILES string

O=C(NC1=NN=CC=C1)N(CC2)CCC2=CC3=CC=CC(OC4=NC=C(C(F)(F)F)C=C4)=C3

InChI

1S/C23H20F3N5O2/c24-23(25,26)18-6-7-21(27-15-18)33-19-4-1-3-17(14-19)13-16-8-11-31(12-9-16)22(32)29-20-5-2-10-28-30-20/h1-7,10,13-15H,8-9,11-12H2,(H,29,30,32)

InChI key

BATCTBJIJJEPHM-UHFFFAOYSA-N

Biochem/physiol Actions

PF-04457845 is a potent, orally active, irreversible inhibitor of fatty acid amide hydrolase (FAAH), the principle enzyme involved in the degradation of the endocannabinoid anandamide. PF-04457845 is a covalent inhibitor that carbamylates FAAH′s serine nucleophile. It was shown to be both potent and selective against other serine hydrolases. It has an IC50 value of 7.2 nM for human FAAH. The endocannabinoid system is a target for therapeutic pain relief. In a rat model of inflammatory pain, PF-04457845 produced significant reduction of inflammatory pain with efficacy comparable to that of naproxen at 10 mg/kg.
Potent irreversible FAAH inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service