Skip to Content
Merck
All Photos(1)

Key Documents

PZ0113

Sigma-Aldrich

PD173952

≥98% (HPLC)

Synonym(s):

6-(2,6-Dichlorophenyl)-8-methyl-2-(4-morpholin-4-ylphenylamino)-8H-pyrido[2,3-d]pyrimidin-7-one, PD-173952

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21Cl2N5O2
CAS Number:
Molecular Weight:
482.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

yellow to green

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

CN1C(=O)C(=Cc2cnc(Nc3ccc(cc3)N4CCOCC4)nc12)c5c(Cl)cccc5Cl

InChI

1S/C24H21Cl2N5O2/c1-30-22-15(13-18(23(30)32)21-19(25)3-2-4-20(21)26)14-27-24(29-22)28-16-5-7-17(8-6-16)31-9-11-33-12-10-31/h2-8,13-14H,9-12H2,1H3,(H,27,28,29)

InChI key

XZEJMVDCQZRHLN-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

PD173952 is a Src family kinase inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Rohe et al.
Bioorganic & medicinal chemistry, 23(15), 4936-4942 (2015-06-11)
Myt1 kinase is a member of the Wee-kinase family involved in G2/M checkpoint regulation of the cell cycle. So far, no peptide substrate suitable for activity-based screening has been reported, hampering systematic development of Myt1 kinase inhibitors. Myt1 inhibitors had

Articles

Src is the original member of a group of non-receptor protein-tyrosine kinases termed the Src family kinases (SFKs). Learn about the SFKs that, in humans, consist of eight members, a small group of atypical members, and two related kinases.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service