Skip to Content
Merck
All Photos(3)

Key Documents

PLA0175

Sigma-Aldrich

Rabbit anti-BRF1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

90kD, 90kD subunit, B - related factor 1, B-related factor 1, BRF, BRF-1, BRF1 homolog, CFDS, GTF3B, GTF3B subunit 2, HEL-S-76p, RNA polymerase III, TAF3B2, TAF3C, TAFIII90, TATA box binding protein (TBP)-associated factor, TATA box binding protein (TBP)-associated factor 3C, TATA box-binding protein-associated factor, TBP - associated factor, TF3B90, TFIIIB90, epididymis secretory sperm binding protein Li 76p, general transcription factor IIIB, hBRF, hTFIIIB90, subunit 2, subunit of RNA polymerase III transcription initiation factor IIIB, subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:200- 1:1,000
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_001510.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... BRF1(2972)

Immunogen

The epitope recognized by PLA0175 maps to a region between residue 627 and 677 of human B - related factor 1 using the numbering given in entry NP_001510.2 (GeneID 2972).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

BRF1 is one of three subunits of the RNA polymerase III transcription factor complex, TFIIIB. TFIIIB regulates RNA Polymerase III transcription which manufactures tRNAs, 5S ribosomal RNA and several other untranslated RNAs.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service