MTCC2000G
MTC™ Bio SureSeal S™ Microcentrifuge Tube
capacity 1.5 mL, sterile, green, pack of 500 ea
Synonym(s):
centrifuge tube, plastic tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.15
Recommended Products
material
conical bottom
green
polypropylene
sterility
sterile
packaging
pack of 500 ea
manufacturer/tradename
MTC™ Bio C2000-G
parameter
28,000 × g speed
capacity
1.5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation, and general sample/reagent storage. SureSeal(TM) S tubes are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity, centrifugal strength (tested up to 28,000×g) and the usable temperature range (-90°C to +121°C). The full-sized tube cap (with frosted marking surface), combined with the unique ring and groove closure, enhances the security of the liquid-tight seal, while minimizing the required locking forces for a more ergonomic opening/closing of the tube caps.
Microtube w/ cap, 1.5ml, green, sterile, w/ self-standing & Stop-Pops™, 500/pk
Features and Benefits
- Sterile comes standard – by electron beam radiation
- Self-Standing bag – with gusseted bottom
- Stop-Pops™ supplied – 10 cap locks in every bag
Legal Information
MTC is a trademark of MTC Bio
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Stop-Pops is a trademark of MTC Bio
SureSeal S is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service