This product is provided as a box of thirteen vials, where each vial contains 10 milligrams of one of thirteen compounds. The compounds are provided as powders, but it appears that all of the compounds are water soluble. The recommendation would be to dissolve them in pure water (not buffer) immediately before use. Solutions may be frozen in aliquots if the entire 10 mg is not going to be used at once.
storage temp.
−20°C
Other Notes
Contains 10 mg each of the urea cycle metabolite standards packaged individually.
For additional information on our range of Biochemicals, please complete this form.
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Kit Components Also Available Separately
Product No.
Description
SDS & Pricing
- L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0% 10 mg
- L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0% 10 mg
See All (13)
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Buenos días. En qué medio se resuspenden estos metabolitos?
1 answer-
Helpful?
-





