MAK516
Myeloperoxidase (MPO) Assay Kit
sufficient for 100 fluorometric tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.84
Recommended Products
Quality Level
detection method
fluorometric
relevant disease(s)
hematological disorder; cancer; cardiovascular diseases
shipped in
wet ice
storage temp.
−20°C
General description
Myeloperoxidase (MPO) is a peroxidase enzyme and can be found in neutrophils, monocytes, and some soft tissue macrophages. MPO has an ability to use chloride as a cosubstrate with hydrogen peroxide to generate hypochlorous acid, a powerful antimicrobial agent produced by neutrophils. However, an excessive production of hypochlorous acid can lead to oxidative stress and tissue damage. Inflammation may also result when MPO oxidizes various substances such as phenols and anilines. Studies show that increased MPO levels may increase the risk of myocardial infarction and cardiovascular disease.
Features and Benefits
- Linear detection range: 0.0025 to 2 U/L
- Compatible with high-throughput handling systems.
Suitability
The kit is suitable for myeloperoxidase (MPO) peroxidation activity determination in biological samples (e.g. cell lysates, tissues, etc.).
Principle
The Myeloperoxidase (MPO) Assay Kit is based on the MPO enzyme reaction with hydrogen peroxide (H2O2) which oxidizes the dye reagent to a highly fluorescent product. The fluorescence intensity of this product, measured at λEx = 530 nm/λEm = 585 nm , is proportional to the total peroxidation activity in the sample. The provided MPO inhibitor is used to suppress peroxidase activity due to MPO in order to differentiate other peroxidase activities that may be present in the sample.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service