M3937
Greiner UV-STAR®384 well plates
cycloolefine, case of 40 ea (internal packs of 10), non-sterile
Synonym(s):
384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
cycloolefine
sterility
non-sterile
feature
lid: no
packaging
case of 40 ea (internal packs of 10)
manufacturer/tradename
Greiner 781801
maximum volume
138 μL
size
384 wells
working volume
110 μL
color
clear
binding type
(Clear film surface treatment)
General description
384 well flat bottom, non-treated microtiter plate with an optimal working volume of 15-110 μL; maximum well capacity of 138 μL.
Other Notes
384 flat well microtiter plate, without lid, rounded square well design, alphanumeric well coding; length: 127.76 +/- 0.15 mm; width: 85.48 +/- 0.15 mm; curvature: approx.150 μm.
Suitability
For μLtra VLT Applications. Highly recommended for measurements of DNA and protein concentrations at 260 nm or 280 nm, respectively. No need for expensive and fragile quartz glass plates. Free of detectable DNase/RNase, human DNA and pyrogens.
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
UV-Star is a registered trademark of Greiner Bio-One GmbH
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service