Skip to Content
Merck

Skip To

M042000

Biotin Phosphoramidite

Synonym(s):

Biotin amidite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C46H64N5O8PS
CAS Number:
Molecular Weight:
878.07
UNSPSC Code:
41116105
PubChem Substance ID:
MDL number:
Assay:
≥96% (31P-NMR)
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Proligo Reagents

Quality Segment

assay

≥96% (31P-NMR)

suitability

conforms to structure for H-NMR

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(N2C3CS[C@@H](CCCCC(=O)NCCOCCOCCOP(OCCC#N)N(C(C)C)C(C)C)C3NC2=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C46H64N5O8PS/c1-34(2)51(35(3)4)60(58-27-12-25-47)59-32-31-57-30-29-56-28-26-48-43(52)16-11-10-15-42-44-41(33-61-42)50(45(53)49-44)46(36-13-8-7-9-14-36,37-17-21-39(54-5)22-18-37)38-19-23-40(55-6)24-20-38/h7-9,13-14,17-24,34-35,41-42,44H,10-12,15-16,26-33H2,1-6H3,(H,48,52)(H,49,53)/t41-,42-,44-,60?/m0/s1

InChI key

WSDQYLGSIUJJRU-ZMKGMYJVSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M042030M042180M042080
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥96% (31P-NMR)

assay

≥96% (31P-NMR)

assay

≥96.0% (reversed phase HPLC), ≥97% (31P-NMR)

assay

≥96% (31P-NMR)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

suitability

conforms to structure for H-NMR

suitability

conforms to structure for H-NMR

suitability

conforms to structure for H-NMR

suitability

conforms to structure for H-NMR

product line

Proligo Reagents

product line

Proligo Reagents

product line

Proligo Reagents

product line

Proligo Reagents


Still not finding the right product?

Explore all of our products under Biotin Phosphoramidite


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters