HS120196
Cardboard Slide Tray
Holds 20 x slides 76x26 mm, red (trim)
All Photos(1)
About This Item
UNSPSC Code:
41123302
NACRES:
NB.22
material
cardboard
red (trim)
feature
Holds 20 x slides 76x26 mm
packaging
case of 72 trays
manufacturer/tradename
Heathrow Scientific HS120196
L × W × H
208 × 341 × 9 mm
Features and Benefits
- Easily organize slides with the index on the front cover that corresponds with the number grid inside
- Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
- Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
- Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
- Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
- Disposable
Choose from one of the most recent versions:
Certificates of Analysis (COA)