Skip to Content
Merck
All Photos(2)

Key Documents

HPA021527

Sigma-Aldrich

Anti-MCM3AP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GANP, Anti-KIAA0572, Anti-Map80, Anti-SAC3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
western blot: 0.04-0.4 μg/mL

immunogen sequence

HEPAEDSDPLSRGDHPPDKRPVRLNRPRGGTLFGRTIQDVFKSNKEVGRLGNKEAKKETGFVESAESDHMAIPGGNQSVLAPSRIPGVNKEEETESREKKEDSLRGTPARQSNRSESTDSLGGLSPSEVTAIQCKNIPDYL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCM3AP(8888)

General description

Minichromosome maintenance complex component 3 associated protein (MCM3AP) is also called as GANP (germinal-center associated nuclear protein). The protein shuttles between cytoplasm and nucleus.

Immunogen

80 kDa MCM3-associated protein recombinant protein epitope signature tag (PrEST)

Application

Anti-MCM3AP antibody produced in rabbit has been used in western blotting and immunoprecipitation of MCM3AP protein from nuclear extract. All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Minichromosome maintenance complex component 3 associated protein (MCM3AP) protein is an acetyl transferase enzyme which inhibits the initiation of DNA replication in a licensing-independent manner. However, this protein will not affect elongation process of DNA replication.
The functional interaction between MCM3AP and glucocorticoid receptor regulates cell proliferation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86517

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shailendra Kumar Singh et al.
Nature communications, 4, 1830-1830 (2013-05-09)
Somatic hypermutation in B cells is initiated by activation-induced cytidine deaminase-catalyzed C→U deamination at immunoglobulin variable regions. Here we investigate the role of the germinal centre-associated nuclear protein (GANP) in enhancing the access of activation-induced cytidine deaminase (AID) to immunoglobulin
Waffa Osman et al.
Biochemical and biophysical research communications, 348(4), 1239-1244 (2006-08-18)
Glucocorticoids are widely used to treat inflammatory diseases but have a number of side effects that partly are connected to inhibition of cell proliferation. Glucocorticoids mediated their action by binding to the glucocorticoid receptor. In the present study, we have
Emma Poole et al.
PloS one, 7(10), e45686-e45686 (2012-10-25)
Like all DNA viruses, human cytomegalovirus (HCMV) infection is known to result in profound effects on host cell cycle. Infection of fibroblasts with HCMV is known to induce an advance in cell cycle through the G(0)-G(1) phase and then a
Yoshinori Takei et al.
The Journal of biological chemistry, 277(45), 43121-43125 (2002-09-13)
Minichromosome maintenance (MCM) proteins are essential components of pre-replication complexes, which limit DNA replication to once per cell cycle. MCM3 acetylating protein, MCM3AP, binds and acetylates MCM3 and inhibits cell cycle progression. In the present study, we examined inhibition of
Martin Kosar et al.
Nature communications, 12(1), 3937-3937 (2021-06-26)
Although human nucleoporin Tpr is frequently deregulated in cancer, its roles are poorly understood. Here we show that Tpr depletion generates transcription-dependent replication stress, DNA breaks, and genomic instability. DNA fiber assays and electron microscopy visualization of replication intermediates show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service