Skip to Content
Merck
All Photos(1)

Key Documents

H1377

Sigma-Aldrich

4-Hydroxy-3-methoxyphenylglycol hemipiperazinium salt

Synonym(s):

3-Methoxy-4-hydroxy-phenylglycol piperazine, MHPG piperazine, MOPEG, MOPEG piperazine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CLP 72,500
25 MG
CLP 141,000
50 MG
CLP 238,000
100 MG
CLP 410,000

CLP 72,500


Estimated to ship onMay 04, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CLP 72,500
25 MG
CLP 141,000
50 MG
CLP 238,000
100 MG
CLP 410,000

About This Item

Empirical Formula (Hill Notation):
C9H12O4 · 0.5C4H10N2
CAS Number:
Molecular Weight:
227.26
Beilstein:
7360817
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CLP 72,500


Estimated to ship onMay 04, 2025


Request a Bulk Order

storage temp.

2-8°C

Quality Level

SMILES string

C1CNCCN1.COc2cc(ccc2O)C(O)CO.COc3cc(ccc3O)C(O)CO

InChI

1S/2C9H12O4.C4H10N2/c2*1-13-9-4-6(8(12)5-10)2-3-7(9)11;1-2-6-4-3-5-1/h2*2-4,8,10-12H,5H2,1H3;5-6H,1-4H2

InChI key

KCECBJRHWSOTMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cerebral norepinephrine metabolite.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Wada et al.
Japanese journal of pharmacology, 78(2), 237-240 (1998-11-26)
To study the circadian rhythm of the monoamine and metabolite contents, cerebrospinal fluid was taken from the cisterna magna through a polyethylene catheter tube every 4 hr in the conscious rat. Norepinephrine and 3-methoxy-4-hydroxy-phenylglycol were found to describe a rhythm
M L Laorden et al.
Life sciences, 66(9), 803-815 (2000-03-04)
Previous report from our laboratory showed that morphine produces a stimulatory effect of hypothalamic noradrenaline (NA) turnover concurrently with enhanced pituitary-adrenal response after its acute injection and during withdrawal. In the present work we have studied the effects of acute
J M Duine et al.
Journal of chromatography. B, Biomedical sciences and applications, 716(1-2), 350-353 (1998-11-21)
A reversed-phase chromatographic method with electrochemical detection was developed for the simultaneous determination of 2,3- and 2,5-dihydroxybenzoates, indicators of in vivo hydroxyl free radical formation, monoamines (NE, DA, 5-HT) and their metabolites (MHPG, DOPAC, HVA, 3MT, 5-HIAA). Linearity was observed

Questions

  1. Can this (MHPG) be stored at -20C

    1 answer
    1. The recommended storage temperature for this product is 2-8°C. In general, powder materials are not affected by freezer storage, however, this item has not been tested under these storage conditions.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service