Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 mL | Check Cart for Availability | CLP 594,000 |
About This Item
NACRES:
NA.76
UNSPSC Code:
12352203
Clone:
-
Species reactivity:
-
Application:
—
Citations:
-
CLP 594,000
Check Cart for Availability
feature
wetted part: no
packaging
pkg of 1 mL
manufacturer/tradename
Cytiva NA933-1ML
color
Brown (light)
storage temp.
2-8°C
General description
ECL™ Human IgG, HRP-linked whole Ab (from sheep). The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2° C to 8° C.
Features and Benefits
- Highly species-specific.
- Optimized for use with our range of ECL™ Detection Reagents.
- Recommended dilutions to minimize background.
Preparation Note
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
ECL is a trademark of Cytiva
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| packaging pkg of 1 mL | packaging pkg of 100 μL | packaging pkg of 1 mL | packaging pkg of 1 mL |
| manufacturer/tradename Cytiva NA933-1ML | manufacturer/tradename Cytiva NA931-100UL | manufacturer/tradename Cytiva NA931-1ML | manufacturer/tradename Cytiva NA934-1ML |
| color Brown (light) | color Brown (light) | color Brown (light) | color Brown (light) |
| feature wetted part: no | feature wetted part: no | feature wetted part: no | feature wetted part: no |
signalword
Warning
hcodes
Storage Class
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

