Skip to Content
Merck
All Photos(1)

Key Documents

EHU013431

Sigma-Aldrich

MISSION® esiRNA

targeting human RAD18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGCAGGGGAGCAGGTTAATGGATAATTTCTTGATCAGAGAAATGAGTGGTTCTACATCAGAGTTGTTGATAAAAGAAAATAAAAGCAAATTCAGCCCTCAAAAAGAGGCGAGCCCTGCTGCAAAGACCAAAGAGACACGTTCTGTAGAAGAGATCGCTCCAGATCCCTCAGAGGCTAAGCGTCCTGAGCCACCCTCGACATCCACTTTGAAACAAGTTACTAAAGTGGATTGTCCTGTTTGCGGGGTTAACATTCCAGAAAGTCACATTAATAAGCATTTAGACAGCTGTTTATCACGCGAAGAGAAGAAGGAAAGCCTCAGAAGTTCTGTTCACAAAAGGAAGCCGCTGCCCAAAACTGTATATAATTTGCTCTCTGATCGTGATTTAAAGAAAAAGCTAAAAGAGCATGGATTATCTATTCAAGGAAATAAACAACAGCTCATTAAAAGGCACCAAGAATTTGTACACATGTACAATGCCCAATGCGATGCTTTGCATCCTAAATCAGCTGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megumi Sasatani et al.
PloS one, 10(2), e0117845-e0117845 (2015-02-13)
The ubiquitin ligase RAD18 is involved in post replication repair pathways via its recruitment to stalled replication forks, and its role in the ubiquitylation of proliferating cell nuclear antigen (PCNA). Recently, it has been reported that RAD18 is also recruited
Chen Xie et al.
Journal of cellular physiology, 234(11), 21100-21112 (2019-05-14)
This study aimed at investigating the role of RAD18 in the regulation of glioblastoma development as well as the underlying mechanisms. The human glioblastoma U251 and U87MG cells were transfected with siRNAs specifically targeting RAD18, and the effects of knockdown
Thomas Göhler et al.
The Journal of cell biology, 192(2), 219-227 (2011-01-19)
DNA polymerase η (polη) belongs to the Y-family of DNA polymerases and facilitates translesion synthesis past UV damage. We show that, after UV irradiation, polη becomes phosphorylated at Ser601 by the ataxia-telangiectasia mutated and Rad3-related (ATR) kinase. DNA damage-induced phosphorylation
Ainhoa Nieto et al.
Cell death and differentiation, 27(4), 1200-1213 (2019-09-12)
Cellular DNA is constantly under threat from internal and external insults, consequently multiple pathways have evolved to maintain chromosomal fidelity. Our previous studies revealed that chronic stress, mediated by continuous stimulation of the β2-adrenergic-βarrestin-1 signaling axis suppresses activity of the
Min Peng et al.
The EMBO journal, 33(15), 1698-1712 (2014-06-27)
Several proteins in the BRCA-Fanconi anemia (FA) pathway, such as FANCJ, BRCA1, and FANCD2, interact with mismatch repair (MMR) pathway factors, but the significance of this link remains unknown. Unlike the BRCA-FA pathway, the MMR pathway is not essential for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service