Skip to Content
Merck
All Photos(1)

Key Documents

D9571

Sigma-Aldrich

Dolasetron mesylate hydrate

≥98% (HPLC), powder

Synonym(s):

Anzemet hydrate, Dolasetron methanesulfonate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O3 · CH4O3S · xH2O
CAS Number:
Molecular Weight:
420.48 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: >30 mg/mL

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

O.CS(O)(=O)=O.O=C1CN2[C@@H]3C[C@H](C[C@H]2CC1C3)OC(=O)c4c[nH]c5ccccc45

InChI

1S/C19H20N2O3.CH4O3S.H2O/c22-18-10-21-12-5-11(18)6-13(21)8-14(7-12)24-19(23)16-9-20-17-4-2-1-3-15(16)17;1-5(2,3)4;/h1-4,9,11-14,20H,5-8,10H2;1H3,(H,2,3,4);1H2/t11?,12-,13+,14+;;

InChI key

QTFFGPOXNNGTGZ-BHSFMSKKSA-N

Biochem/physiol Actions

Dolasetron mesylate hydrate is a highly specific and selective serotonin 5-HT3 receptor antagonist, not shown to have activity at other known serotonin receptors and with low affinity for dopamine receptors.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren Trepanier
Journal of feline medicine and surgery, 12(3), 225-230 (2010-03-03)
The control of nausea and vomiting in cats is important in order to prevent the development of food aversion, anorexia (with its associated complications of weight loss and dehydration), and hepatic lipidosis. There are several antiemetic drugs that are clinically
Mehdi Hamadani et al.
Journal of oncology pharmacy practice : official publication of the International Society of Oncology Pharmacy Practitioners, 13(2), 69-75 (2007-09-18)
The last decade has witnessed the great impact of 5-hydroxytryptamine-3 receptor (5-HT(3))antagonists in revolutionizing the management of platinum-based chemotherapy-induced acute nausea and vomiting (CINV). However, despite the availability of a variety of 5-HT(3) antagonists, little data is published to support
Edward B Silberstein
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 49(4), 546-549 (2008-03-18)
The goal of this study was to reduce the salivary symptoms of pain and xerostomia caused by 131I therapy for papillary and follicular thyroid carcinoma. In a single-blind controlled prospective study of 60 patients, we investigated whether pilocarpine, 5 mg
[Pharmacokinetic and pharmacodynamic features of palonosetron].
Armando A Genazzani et al.
Tumori, 94(2), suppl 6-suppl13 (2008-06-21)
Christopher M Peters et al.
Brain research, 1352, 83-93 (2010-07-20)
The balance between descending inhibition and facilitation is thought to be disturbed in chronic pain states. Increased facilitation by spinally released serotonin has been suggested by demonstration that mechanically evoked neuronal responses of wide dynamic range neurons are inhibited by

Articles

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service