Skip to Content
Merck
All Photos(1)

Key Documents

D9069

Sigma-Aldrich

N-Desmethyltamoxifen HCl

≥98% (HPLC), solid

Synonym(s):

(Z)-2-(p-(1,2-Diphenyl-1-butenyl)phenoxy)-N-methylethylamine, ICI 55,548

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27NO·HCl
CAS Number:
Molecular Weight:
393.95
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77

sterility

non-sterile

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

technique(s)

inhibition assay: suitable

solubility

DMSO: ≥20 mg/mL

shipped in

ambient

storage temp.

room temp

SMILES string

Cl.CC\C(c1ccccc1)=C(/c2ccccc2)c3ccc(OCCNC)cc3

InChI

1S/C25H27NO.ClH/c1-3-24(20-10-6-4-7-11-20)25(21-12-8-5-9-13-21)22-14-16-23(17-15-22)27-19-18-26-2;/h4-17,26H,3,18-19H2,1-2H3;1H/b25-24-;

InChI key

GMLHJWZEYLTNJW-BJFQDICYSA-N

Application

N-Desmethyltamoxifen HCl has been used as an internal standard in high-performance liquid chromatography (HPLC) for the quantification of metabolites from NG2 cell-derived astrocytes.

Biochem/physiol Actions

N-Desmethyltamoxifen HCl is the primary metabolite of tamoxifen. N-Desmethyltamoxifen HCl is an estrogen response modifer and protein kinase C inhibitor.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) and PKC pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Injury type-dependent differentiation of NG2 glia into heterogeneous astrocytes
Hackett AR, et al.
Experimental Neurobiology, 308, 72-79 (2018)
Altin Sula et al.
Molecular cell, 81(6), 1160-1169 (2021-01-28)
Voltage-gated sodium channels are targets for many analgesic and antiepileptic drugs whose therapeutic mechanisms and binding sites have been well characterized. We describe the identification of a previously unidentified receptor site within the NavMs voltage-gated sodium channel. Tamoxifen, an estrogen

Articles

Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service