Skip to Content
Merck

D2272

Dialysis tubing, benzoylated

Avg. flat width 9 mm (0.35 in.), MWCO 2000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41123100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

regenerated cellulose

capacity

~10 mL/ft

pore size

2000 MWCO

storage temp.

2-8°C

Application

Cellulose tubing, benzoylated to reduce pore size. Useful for separating compounds with a molecular weight of ≤1,200 from compounds with a molecular weight >2,000.

Packaging

Supplied in a water pack containing 0.002% 1-hydroxypyridine-2-thione as preservative.


Still not finding the right product?

Explore all of our products under Dialysis tubing, benzoylated


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library