CLS7369
Corning® HTS Transwell® 96 well plate
0.4 μm, PET, with barcode, lid, sterile, pkg of 5 ea
Synonym(s):
cell culture plate, multi well plates, multiple well plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
size:
96 wells
sterility
sterile
feature
lid
packaging
pkg of 5 ea
manufacturer/tradename
Corning 7369
size
96 wells
wells
96 wells
suitability
suitable for (cell cuture)
Related Categories
General description
The HTS Transwell-96 permeable support contains Transwell® unit (1 integral tray with 96 wells), reservoir plate (for cell growth steps), receiver plate (96 wells for growth or assay steps) and 2 sterile lids; 5 sets per case
Application
Corning® HTS Transwell® 96 well plate is suitable for air-liquid interface (ALI) cultures.
Features and Benefits
- 0.4 μm pore size polyester (PET) membrane, which is surface treated for optimal cell attachment
- Surface area for Cell growth is 0.143 cm² which is 20-50% greater than other commercially available brands.
- Large apical and basolateral access ports to enable efficient media sampling and facilitate automated or manual access.
- Optimized for automation, with multichannel feeder ports, improved gripping surface, and bar coding.
- The untreated reservoir plate permits simultaneous feeding of 96 wells and it contains a removable media stabilizer to prevent spills during handling.
- The untreated receiver plate isolates each well to enable 96 individual assays.
Legal Information
Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service