CLS4519162
Corning® ProCulture® spinner flask accessories
side arm fitting, Combination with 1/4 inlets for 1 L spinner flask, Gas and Media Handling Fitting with Dual 1/4 in. Inlets (1 long, 1 short tube) for 1L Angled Sidearm Flasks
Synonym(s):
container accessory
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Corning 4519-162
Related Categories
General description
Side arm fittings used for a variety of functions in angled sidearm Spinner Flasks during cell culture. Sidearm fittings provide a convenient way of transferring media into or out of flasks aseptically. They can also be used to sparge the cell culture medium with gases to enhance cell production. Fittings comprise of a PET insert with a Viton O-ring and a polypropylene sealing cap & 316 stainless steel tubes.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Legal Information
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries