Skip to Content
Merck
All Photos(1)

Key Documents

CLS431720

Corning® mini bioreactor

centrifuge tube with vent cap, volume 50 mL, sterile

Synonym(s):

Corning bioreactor, Mini bioreactor, Small-scale bioreactor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122105
NACRES:
NB.43

material

clear tube
conical tube (threaded)
neck (wide)
orange polypropylene cap (vented)
polypropylene tube

sterility

sterile

feature

RCF 12,000 × g
RNase and DNase free
disposable
graduations 5 ‑ 50 mL
self-standing: no

packaging

pack of 25 ea
case of 300 ea

manufacturer/tradename

Corning 431720

O.D. × L

29.1 mm × 114.4 mm

cap diam.

35.2 mm

volume

50 mL

Application

Corning® mini bioreactor is suitable for the following applications:
  • Cell line development
  • Clone selection
  • Media optimization
  • Recombinant protein development

Features and Benefits

  • Polypropylene tube with large marking spot to clearly identify tube contents and experimental parameters
  • Polypropylene cap with 4 vents; hydrophobic membrane provides gas exchange
  • Sterile, nonpyrogenic and RNase-/DNase-free
  • Ideal for high throughput process optimization for suspension cell culture

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service