CLS3640
Corning® NBS™ 384 well microplates
surface treatment, Non-binding Surface, clear polystyrene, flat bottom, non-sterile, lid: no, pack of 25
Synonym(s):
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates
Select a Size
CLP 470,000
Select a Size
About This Item
flat bottom
flat bottom wells clear wells
polystyrene
square clear wells
CLP 470,000
Recommended Products
material
clear polystyrene
flat bottom
flat bottom wells clear wells
polystyrene
square clear wells
sterility
non-sterile
non-sterile
feature
lid: no
lid: no
skirt
plate format: 384 well standard
packaging
case of 100
pack of 25
manufacturer/tradename
Corning 3640
maximum volume
112 μL
size
384 wells
working volume
80 μL
suitability
suitable for (cell cuture)
binding type
NBS™ (non-binding surface)
Related Categories
General description
- Nonbinding (NBS) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
- Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)