Skip to Content
Merck
All Photos(1)

Key Documents

CLS3337

Corning® CellBIND® Multiple Well Plate

size 24 wells, flat clear bottom, sterile, lid

Synonym(s):

24 well culture plate, cell culture multiwell plates, cell culture plates

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
CLP 309,000

CLP 309,000


Estimated to ship onMay 17, 2025



Select a Size

Change View
50 EA
CLP 309,000

About This Item

UNSPSC Code:
41122107
eCl@ss:
32190102
NACRES:
NB.13
material:
clear polystyrene plate
flat bottom wells clear
flat clear bottom
polystyrene
size:
24 wells
sterility:
sterile
sterile
binding type:
CellBIND® surface
feature:
lid
lid
skirt
plate format: 24 well standard

CLP 309,000


Estimated to ship onMay 17, 2025


material

clear polystyrene plate
flat bottom wells clear
flat clear bottom
polystyrene

sterility

sterile
sterile

feature

lid
lid
skirt
plate format: 24 well standard

packaging

pack of 5 ea
case of 50 ea

manufacturer/tradename

Corning 3337

size

24 wells

surface area

1.9 cm2 , cell growth area

well volume

3.4 mL

well working volume

0.38-0.57 mL

color

clear

suitability

suitable for (cell culture involving anchorage-dependant cell lines)

binding type

CellBIND® surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® CellBIND® Cell Culture Multiwell Plates

  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumerical codes for well identification
  • Uniform footprint for ease in stacking
  • CellBIND® surface treated for optimal cell attachment
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhonda Flores et al.
Biomolecules, 9(12) (2019-11-30)
Lecithin:cholesterol acyltransferase (LCAT) is an enzyme secreted by the liver and circulates with high-density lipoprotein (HDL) in the blood. The enzyme esterifies plasma cholesterol and increases the capacity of HDL to carry and potentially remove cholesterol from tissues. Cholesterol accumulates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service