Skip to Content
Merck
All Photos(1)

Documents

P19 Cell Line from mouse

95102107, mouse embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

P19 Cell Line from mouse, 95102107

biological source

mouse embryo

description

Mouse teratocarcinoma

growth mode

Adherent

karyotype

Euploid

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse teratocarcinoma

Cell Line Description

The P19 cell line was derived from an embryonal carcinoma induced in a C3H/He strain mouse. The cells can be cloned at high efficiency in medium containing 0.1mM β-mercaptoethanol. The pluripotent P19 cells can be induced to differentiate into neuronal and glial cells in the presence of retinoic acid. Aggregates of P19 cells differentiate into cardiac and skeletal muscle in the presence of dimethyl sulfoxide (DMSO). In the presence of both retinoic acid and DMSO cells develop as if exposed to retinoic acid only.

Application

Differentiation studies

Culture Medium

α MEM + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 2.5% Foetal Bovine Serum (FBS) + 7.5% Calf Serum (CS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service