Skip to Content
Merck

L5178Y

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
mouse thymus
Relevant disease(s):
cancer
Growth mode:
Suspension
Karyotype:
Not specified
Morphology:
Lymphoblast
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse thymus

packaging

tube of 5 μg 87111908-DNA-5UG, pkg of vial of cells 87111908-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

Toxicity screening and radiation studies

Biochem/physiol Actions

Derived from a thymic tumour induced in a DBA/2 mouse by methylcholanthrene. The cells were adapted for in vitro growth. The cells have been widely used in toxicity and radiation studies.
Mouse DBA/2 lymphoma

Preparation Note

Fischer′s + 2mM Glutamine + 1% Sodium Pyruvate (NaP) + 10% Horse Serum (HS) or RPMI 1640 + 10% Foetal Bovine Serum (FBS).
Maintain cultures between 3x10,000 to 5x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to [email protected].




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library