Skip to Content
Merck
All Photos(1)

Key Documents

OX-106

13082220, from Mouse x Mouse Hybridoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

Rat leukocytes

Myeloma

NS1

Reactivity

Rat neutrophils

technique(s)

cell culture | mammalian: suitable

isotype

Not specified

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

The hybridoma cell line OX-106 secretes monoclonal antibodies reactive with rat neutrophils.

Immunogen

Rat leukocytes

Application

Neutrophil recognition.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Seed at 5 x 104 cells/ml (5 x 105 cells/ml at resuscitation). Subculture when cells reach 5 x 105 - 1 x 106 cells/ml (usually after 3-4 days), 5% CO2, 37°C. 20% FBS can be used at resuscitation.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service