Skip to Content
Merck
All Photos(1)

Key Documents

COV413B

7071906, human ovary (metastasis), Polygonal & fusiform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

COV413B, 07071906

biological source

human ovary (metastasis)

growth mode

Adherent

karyotype

Modal chromosome no. 38 (36-42)

morphology

Polygonal & fusiform

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ovarian epithelial-serous carcinoma, bladder dome

Cell Line Description

A human ovarian epithelial-serous carcinoma cell line established from an intraperitoneal metastasis of the bladder dome. This cell line was derived from the same patient as the ovarian cell line COV413A (ECACC cat no. 07071905). Anchorage-independent growth in agar has been reported.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service