Skip to Content
Merck
All Photos(1)

Key Documents

211F-500

Sigma-Aldrich

Endothelial Cell Growth Medium without FBS (500ml)

Synonym(s):

Endothelial Cell Culture Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media
Endothelial Cell Growth Medium without FBS (500ml) is an all-in-one ready-to-use media. The complete Growth Medium is optimized and best-suited for angiogenesis and other physiological studies.

Application

Endothelial Cell Growth Medium without FBS (500ml) has been used for culturing human aortic endothelial cells and human umbilical vein endothelial cells.

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Wang et al.
Die Pharmazie, 71(10), 592-597 (2016-10-01)
Human abdominal aortic aneurysm (AAA) is characterized by the induction of intracellular and extracellular inflammatory cytokines and the production of reactive oxygen species (ROS) associated with localized inflammatory responses in the vascular wall. Recent studies have shown that greater circulating
Chin-Feng Hsuan et al.
Molecules (Basel, Switzerland), 20(9), 16908-16923 (2015-09-24)
Chronic inflammation plays a pivotal role in the development of atherosclerosis, where the pro-inflammatory cytokine-induced expression of endothelial adhesion molecules and the recruitment of monocytes are the crucial events leading to its pathogenesis. Glossogyne tenuifolia ethanol extract (GTE) is shown

Protocols

Discover our collection of primary human hepatic sinusoidal endothelial cells, a protocol for thawing, plating, and growing HHSECs, and a recipe for our HHSEC maintenance media formulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service