Skip to Content
Merck
All Photos(1)

Key Documents

NIST2585

Organic contaminants in house dust

NIST® SRM® 2585

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 10 g

manufacturer/tradename

NIST®

application(s)

industrial hygiene

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in evaluating analytical methods for the determination of selected polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, and polybrominated diphenyl ether (PBDE) congeners in house dust and similar matrices. All of the constituents for which certified, reference, and information values are provided in SRM 2585 were naturally present in the dust material before processing. A unit of SRM 2585 consists of one bottle containing approximately 10 g of house dust. For more information, please refer to the COA and SDS.

SRM 2585_cert

SRM 2585_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Lact. - Repr. 1B - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service