Skip to Content
Merck
All Photos(1)

Key Documents

NIST2034

Holmium Oxide Solution Wavelength Standard (240 nm to 650 nm)

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 6 g

manufacturer/tradename

NIST®

application(s)

general analytical

format

single component solution

storage temp.

15-25°C

General description

Each unit of SRM 2034 contains a solution in a flame-sealed, nonfluorescent, fused-silica cuvette of optical quality (parallel to ∼0.9 mrad and flat to ∼1 µm). For more information, please refer to the COA and SDS.


SRM 2034_cert

SRM 2034_SDS

Features and Benefits

  • The certified wavelengths of minimum transmittance are provided for 14 bands ranging from 240 nm to 650 nm and six spectral bandwidths from 0.1 nm to 3.0 nm.
  • Informational values for the prediction intervals of the certified wavelengths are provided at an approximate 95% confidence level.
  • Accompanied by instructions for handling, storage, and use, along with maintenance and expiration details of the SRM certificate.

Other Notes

Certified for the analytes listed below.
Holmium oxide (Ho2O3)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Ox. Liq. 2 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service