Skip to Content
Merck
All Photos(5)

Key Documents

CRM00506L

Millipore

Paraclostridium bifermentans (former Clostridium bifermentans) NCTC 506 Low count LENTICULE® discs

greener alternative

Clostridium bifermentans NCTC 506 LENTICULE® discs, 30-120 CFU, suitable for microbiology, certified reference material

Synonym(s):

C. bifermentans NCTC 506 Low count Lenticule® discs, Certified reference material, Microbiological standards

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

ISO 17034
ISO/IEC 17025

form

disc (hydrosoluble)
solid

shelf life

limited shelf life, expiry date on the label

packaging

pack of 10

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

sustainability

Greener Alternative Product

application(s)

environmental
food and beverages

microbiology

greener alternative category

storage temp.

−20°C

suitability

Clostridium bifermentans
Clostridium spp.

General description

Clostridium bifermentans NCTC 506 Low count Lenticule® discs are Certified Microorganisms Standards (Certified Microbiological Reference Materials) which can be used as quantitative and qualitative control organisms. They contain colony forming units (CFUs) in the range of 30-120 CFU per disc. The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms. These microorganisms are certified according to ISO/IEC 17025 and produced under reproducible conditions compliant with ISO 17034 using authenticated strains from NCTC. These ready-to-use microbiological controls minimize the need to maintain control strains in the test laboratory and guarantee an authenticated control culture for every quality control test. Lenticule® discs are prepared from a traceable culture obtained freeze-dried from the National Collection of Type Cultures (NCTC) or National Collection of Pathogenic Fungi (NCPF) and are manufactured by Sigma-Aldrich under license from UK Health Security Agency.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of our four categories of greener alternatives. Click here to view its DfS scorecard.

Application

Clostridium bifermentans NCTC 506 Low count Lenticule® discs are used as reference strains in routine quality control for food, beverage, water, and environmental testing. Additionally, they are excellent for use in method development and validations, as starter cultures, and for use in research studies and educational purposes.

Features and Benefits

  • Ready-to-use at convenient CFU ranges
  • No dilution required
  • Quick rehydration in 10 minutes
  • At least 16 months of shelf life in a standard -20 °C freezer
  • Complete with a comprehensive certificate of analysis
  • No maintenance of stock cultures
  • No recovery time or pre-enrichment step is necessary
  • Robust, efficient, and reliable
  • Easy-to-use
  • Cost-effective

Other Notes

Strain origin: NCTC 506

Other cell culture collections:

  • DSM 103419
  • CECT 550
Click here to access the BacDive database for more info on metabolism, genome, growth media, and more
Strain History - Accessioned into the NCTC in 1920 < M. Robertson (Lister Institute of Preventative Medicine, London, UK). Isolation substrate unknown, geographic orgin unknown, before 1920.

Legal Information

Lenticule is a registered trademark of The Secretary of State for Health (United Kingdom)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service