Skip to Content
Merck
All Photos(1)

Key Documents

COND1412

Supelco

Conductivity Standard - 1412 μmhos/cm

certified reference material

Synonym(s):

1412 μmhos/cm Conductivity Calibration Standard

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
CLP 67,700
1 L
CLP 90,200

CLP 67,700


In StockDetails


Request a Bulk Order

Select a Size

Change View
500 ML
CLP 67,700
1 L
CLP 90,200

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CLP 67,700


In StockDetails


Request a Bulk Order

grade

certified reference material

Quality Level

100
300

CofA

current certificate can be downloaded

manufacturer/tradename

RTC COND-1412-1

concentration

in H2O

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This certified reference material (CRM) is designed, prepared and qualified for accuracy and stability according to ISO 17034 and ISO/IEC 17025. The specific conductance value of 1412 ± 6 µmhos/ cm is quantified using SI and higher order standards from NIST through an unbroken chain of comparisons.

The certificate of analysis (CoA) includes information such as traceability, uncertainty, stability, homogeneity and safety.

Application

Used for conductivity calibration verification.

Featured as a standard in the fabrication of electrical conductivity sensors for 2D mapping of ammonia released into the water.[1]

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Developing a network of inexpensive electrical conductivity sensors to map ammonia release
Collins MD, et al.
Chemical, Biological, Radiological, Nuclear, and Explosives (CBRNE) Sensing XIII, 12116, 133-139 (2022)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service