Skip to Content
Merck
All Photos(1)

Documents

C2177000

Cinchocaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dibucaine hydrochloride, 2-Butoxy-N-(2-diethylaminoethyl)-4-quinolinecarboxamide hydrochloride, Cinchocaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29N3O2 · HCl
CAS Number:
Molecular Weight:
379.92
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cinchocaine

manufacturer/tradename

EDQM

mp

99-101 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CCCCOc1cc(C(=O)NCCN(CC)CC)c2ccccc2n1

InChI

1S/C20H29N3O2.ClH/c1-4-7-14-25-19-15-17(16-10-8-9-11-18(16)22-19)20(24)21-12-13-23(5-2)6-3;/h8-11,15H,4-7,12-14H2,1-3H3,(H,21,24);1H

InChI key

IVHBBMHQKZBJEU-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Cinchocaine hydrochloride is a drug commonly used in ophthalmic ointments to produce conjunctival anaesthesia or in dilute penetrable solutions to produce spinal or surface anesthesia.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Potent, long-acting local anesthetic

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of chlorpromazine and thioridazine hydrochlorides in pharmaceutical formulations.
Mehta A C.
Analyst, 106(1267), 1119-1122 (1981)
Cinchocaine hydrochloride determination by atomic absorption spectrometry and spectrophotometry.
Abdel-Ghani NT, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 60(5), 419-424 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service