Skip to Content
Merck
All Photos(2)

Key Documents

73177

Supelco

1-Fluoro-2,4-dinitrobenzene

for HPLC derivatization, LiChropur, ≥99.0% (GC)

Synonym(s):

2,4-Dinitro-1-fluorobenzene, DNFB, DNPF, FDNB, Sanger reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3F
CAS Number:
Molecular Weight:
186.10
Beilstein:
398632
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥99.0% (GC)

form

(Solid or Semi-Solid or Chunk(s) or Paste or Liquid)

quality

LiChropur

technique(s)

HPLC: suitable

color

light yellow to very dark green-yellow, and Light Green to Very Dark Green

refractive index

n20/D 1.569 (lit.)

bp

178 °C/25 mmHg (lit.)

mp

25-27 °C (lit.)

density

1.482 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(F)c(c1)[N+]([O-])=O

InChI

1S/C6H3FN2O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H

InChI key

LOTKRQAVGJMPNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Fluoro-2,4-dinitrobenzene is a color labeling agent, mainly used for detecting primary and secondary amines.

Application

1-Fluoro-2,4-dinitrobenzene may be used as a derivatizing reagent for the determination of gabapentin in human plasma samples, pharmaceutical formulations and memantine hydrochloride in pharmaceutical formulations using high-performance liquid chromatography technique.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

327.2 °F

Flash Point(C)

164 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of an HPLC method for determination of gabapentin in dosage forms through derivatization with 1-fluoro-2, 4-dinitrobenzene
Souri E, et al.
Chemical & Pharmaceutical Bulletin, 55(10), 1427-1430 (2007)
A stability-indicating HPLC method for the determination of memantine hydrochloride in dosage forms through derivatization with 1-fluoro-2, 4-dinitrobenzene
Jalalizadeh H, et al.
Scientia Pharmaceutica, 82(2), 265-280 (2014)
Validated HPLC method for the determination of gabapentin in human plasma using pre-column derivatization with 1-fluoro-2, 4-dinitrobenzene and its application to a pharmacokinetic study
Jalalizadeh H, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 854(1-2), 43-47 (2007)
Sonia Schuepbach-Mallepell et al.
The Journal of allergy and clinical immunology, 132(6), 1348-1357 (2013-08-21)
Innate immune sensors control key cytokines that regulate T-cell priming and T-cell fate. This is particularly evident in allergic reactions, which represent ideal systems to study the interplay of innate and adaptive immunity. In patients with contact dermatitis, inflammasome-mediated IL-1
Morten M Nielsen et al.
Journal of immunology (Baltimore, Md. : 1950), 192(7), 2975-2983 (2014-03-07)
Substances that penetrate the skin surface can act as allergens and induce a T cell-mediated inflammatory skin disease called contact hypersensitivity (CHS). IL-17 is a key cytokine in CHS and was originally thought to be produced solely by CD4(+) T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service