Skip to Content
Merck
All Photos(2)

Documents

68300

Supelco

Tazobactam sodium salt

analytical standard

Synonym(s):

CL-298741, YTR-830H, [2S-(2α, 3β, 5α)]-3-Methyl-7-oxo-3(1H,2,3-triazol-1-ylmethyl)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N4NaO5S
CAS Number:
Molecular Weight:
322.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]/D 144±4°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].C[C@]2(Cn1ccnn1)[C@@H](N3[C@@H](CC3=O)S2(=O)=O)C([O-])=O

InChI

1S/C10H12N4O5S.Na/c1-10(5-13-3-2-11-12-13)8(9(16)17)14-6(15)4-7(14)20(10,18)19;/h2-3,7-8H,4-5H2,1H3,(H,16,17);/q;+1/p-1/t7-,8+,10+;/m1./s1

InChI key

RFMIKMMOLPNEDG-QVUDESDKSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tazobactam sodium is the sodium salt of the β-lactamase inhibitor antibiotic, tazobactam. It is a semisynthetic parenteral penicillin which can be combined with piperacillin and ceftolozane drugs for treating a variety of infections.

Application

Tazobactam sodium may be used as an analytical reference standard for the determination of the analyte in pharmaceutical formulations by reversed-phase high-performance liquid chromatography.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RP-HPLC method for simultaneous estimation of cefepime hydrochloride and tazobactam sodium in bulk and pharmaceuticals
Tamboli SR and Patil DD
Journal of Chemistry, 2013(4), 543-543 (2013)
Stability indicating RP-HPLC method for the estimation of ceftazidime pentahydrate and tazobactam sodium in bulk and dosage forms
Amareswari S, et al.
Indian Journal of Pharmaceutical and Biological Research, 1(4), 543-543 (2013)
Simultaneous determination of piperacillin and tazobactam in pharmaceutical formulations by RP-HPLC method
Veni KRP, et al.
Journal of Pharmacy Research, 7(1), 127-131 (2013)
RP-HPLC method for simultaneous estimation of cefepime hydrochloride and tazobactam sodium in bulk and pharmaceuticals
Tamboli SR and Patil DD
Journal of Chemistry, 2013(1), 127-131 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service