Skip to Content
Merck
All Photos(2)

Key Documents

56601

Supelco

Icariin

analytical standard

Synonym(s):

4′-O-methyl-8-γ,γ-dimethylallylkaempferol-3-rhamnoside-7-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40O15
CAS Number:
Molecular Weight:
676.66
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2=C(O[C@@H]3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)C(=O)c4c(O)cc(O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)c(C\C=C(\C)C)c4O2

InChI

1S/C33H40O15/c1-13(2)5-10-17-19(45-33-28(42)26(40)23(37)20(12-34)46-33)11-18(35)21-24(38)31(48-32-27(41)25(39)22(36)14(3)44-32)29(47-30(17)21)15-6-8-16(43-4)9-7-15/h5-9,11,14,20,22-23,25-28,32-37,39-42H,10,12H2,1-4H3/t14-,20+,22-,23+,25+,26-,27+,28+,32-,33+/m0/s1

InChI key

TZJALUIVHRYQQB-XLRXWWTNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Icariin is a major flavonoid isolated from Epimedium spp. leaves, used as a traditional Chinese medicine to nourish the kidney and reinforce yang.

Application

Icariin has been used as a test material to investigate its, in vitro effect in promoting mouse hair follicle growth, which is assessed by vibrissae hair follicle (VHF) organ-culture model.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Flavonoid glucoside derived from the the Epimedium plant genus. This pharmacologically active constituent or its derivatives have been shown to enhance bone formation, modulate multidrug resistance, enhance neuronal viability, stimulate angiogenesis, and exhibit antioxidant activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Icariin promotes mouse hair follicle growth by increasing insulin?like growth factor 1 expression in dermal papillary cells.
Su SY, et al.
Clinical and Experimental Dermatology, 42(3), 287-294 (2017)
Transforming Growth Factors?Advances in Research and Application (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service