Skip to Content
Merck
All Photos(1)

Documents

33627

Supelco

Alkylphenol Internal Standard Mix 7-solution for DIN EN ISO 18857-2

in acetone, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Agency

DIN
EN

shelf life

limited shelf life, expiry date on the label

concentration

in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

−20°C

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

10 mL solution in acetone contain:
4-tert-Octylphenol (ring 13C6) 1 μg/mL
4-tert-Octylphenol monoethoxylate (ring 13C6) 1 μg/mL
4-tert-Octylphenol diethoxylate (ring 13C6) 1 μg/mL
4-(3,6-Dimethyl-3-heptyl)-phenol (ring 13C6) 1 μg/mL
4-(3,6-Dimethyl-3-heptyl)-phenol monoethoxylate (ring 13C6) 1 μg/mL
4-(3,6-Dimethyl-3-heptyl)-phenol diethoxylate (ring 13C6) 1 μg/mL
Bisphenol A (D16) 1 μg/mL

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service