Skip to Content
Merck
All Photos(1)

Documents

33034

Sigma-Aldrich

L-Ascorbic acid

puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

ACS reagent
puriss. p.a.

Agency

Ph. Eur.
USP/NF
reag. ISO

Assay

99.7-100.5% (oxidimetric)

form

solid

optical activity

[α]20/D +20.5 to +21.5°, c = 10% in H2O

technique(s)

IR spectroscopy: suitable

impurities

related substances, in accordance (HPLC)
≤0.001% heavy metals (by ICP-OES)

ign. residue

≤0.05% (as SO4)

loss

≤0.1% loss on drying, 105 °C

pH

2.1-2.6 (20 °C, 5%)

mp

190-192 °C ((decomp)) (lit.)
190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

As: ≤1 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution
passes test for identity (IR)

application(s)

cell analysis

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid or vitamin C is a water-soluble vitamin found in various foodstuffs. Benefits of vitamin C may include protection against immune system deficiencies, cardiovascular disease, hyperpigmentation, skin protection, anti-aging, iron absorption, and red blood cell production. It is also useful in the biosynthesis of collagen, L-carnitine, and neurotransmitters.

Application

L-Ascorbic acid has been used as an analytical interfering substance in glycated albumin (GA) enzymatic assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin C: One compound, several uses. Advances for delivery, efficiency and stability
Testa R, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 1(2), 162-171 (2016)
T R Tickner et al.
Clinica chimica acta; international journal of clinical chemistry, 85(2), 125-129 (1978-04-17)
A new colorimetric method for the assay of biliverdin in biological fluids is described. The method, based upon the reaction of biliverdin with barbituric acid, offers improved sensitivity and selectivity when compared to direct spectrophotometric measurements. Using this method biliverdinaemia
Total phenols, ascorbic acid, β-carotene and lycopene in Portuguese wild edible mushrooms and their antioxidant activities.
Barros L, et al.
Food Chemistry, 103(2), 413-419 (2007)
A family of mammalian NaI[plus]I-dependent L-ascorbic acid transporters.
Tsukaguchi H, et al.
Nature, 673(1), 70-75 (1999)
Vitamin C: One compound, several uses. Advances for delivery, efficiency and stability
Carita AC, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 31(2), 102117-102117 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service