20780-U
Hamilton® GASTIGHT® syringe, 1700 series, removable needle
1701RN, volume 10 μL, needle size 26s ga (bevel tip), needle L 51 mm (2 in.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
glass body
needle size
26s ga (bevel tip)
feature
removable needle RN
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80030
needle L
51 mm (2 in.)
volume
10 μL
Looking for similar products? Visit Product Comparison Guide
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service