Skip to Content
Merck
All Photos(1)

Key Documents

19078

Supelco

Platinum Standard for ICP

TraceCERT®, 1 g/L Pt in hydrochloric acid (nominal concentration)

Synonym(s):

ICP Platinum Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Pt in hydrochloric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Platinum standard may have been used as reference standard in laser ablation inductively coupled plasma mass spectrometry (LA-ICP-MS) method for determination of trace elements in high-purity platinum and trace levels of platinum in peripheral nerves.

Analysis Note

1 g/L Pt in 5% hydrochloric acid (contains HNO3 traces), prepared with high purity Pt metal, HCl, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of an ICP-MS assay for the detection of trace levels of platinum in peripheral nerves.
Screnci, D., et al.
Atomic Spectroscopy, 19, 172-175 (1998)
Determination of trace elements in high-purity platinum by laser ablation inductively coupled plasma mass spectrometry using solution calibration.
Becker, J. Sabine; Pickhardt, Carola, and Dietze, Hans-Joachim.
Journal of Analytical Atomic Spectrometry, 16.6, 603-606 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service