Skip to Content
Merck
All Photos(1)

Key Documents

09774

Sigma-Aldrich

Fmoc-Bpa-OH

≥98.0%

Synonym(s):

Fmoc-4-benzoyl-L-phenylalanine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CLP 364,000

CLP 364,000


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
1 G
CLP 364,000

About This Item

Empirical Formula (Hill Notation):
C31H25NO5
CAS Number:
Molecular Weight:
491.53
Beilstein:
7672022
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

CLP 364,000


Estimated to ship onMay 17, 2025


Request a Bulk Order

Quality Level

Assay

≥98.0%

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccc(cc1)C(=O)c2ccccc2)NC(=O)OCC3c4ccccc4-c5ccccc35

InChI

1S/C31H25NO5/c33-29(21-8-2-1-3-9-21)22-16-14-20(15-17-22)18-28(30(34)35)32-31(36)37-19-27-25-12-6-4-10-23(25)24-11-5-7-13-26(24)27/h1-17,27-28H,18-19H2,(H,32,36)(H,34,35)/t28-/m0/s1

InChI key

SYOBJKCXNRQOGA-NDEPHWFRSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Zhi Zhao et al.
Molecules (Basel, Switzerland), 23(8) (2018-07-28)
HIV-1 integrase (IN) inhibitors represent a new class of highly effective anti-AIDS therapeutics. Current FDA-approved IN strand transfer inhibitors (INSTIs) share a common mechanism of action that involves chelation of catalytic divalent metal ions. However, the emergence of IN mutants

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service