11699075001
Roche
Nylon Membranes for Colony and Plaque Hybridization
Synonym(s):
membrane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
packaging
pkg of 50 discs (82 mm diameter)
Quality Level
manufacturer/tradename
Roche
disc diam.
82 mm
storage temp.
20-25°C
General description
Nylon membrane is a circular, uncharged, microporous, amphoteric, ultra-pure nylon 66 membrane on a polyester support. It has a durable structure with consistent transfer rate.
Surface Properties: Pore size of 1.2 μm. Surface contains 50% amino and 50% carboxyl groups.
Surface Properties: Pore size of 1.2 μm. Surface contains 50% amino and 50% carboxyl groups.
Application
- The membrane discs are specifically suited for:Nonradioactive screening of phage or cosmid libraries with digoxigenin-labeled probes followed by detection with highly sensitive chemiluminescent or chromogenic substrates
- Hybridization with biotinylated or radioactive probes
- It has been used in the colony immunoblotting technique.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
K P Karey et al.
Analytical biochemistry, 178(2), 255-259 (1989-05-01)
Western blotting of low molecular weight (Mr) acidic and basic isoelectric point (pI) proteins was studied to optimize detection sensitivities. Radioiodinated epidermal growth factor (EGF, Mr 6045, pI 4.4), transforming growth factor type alpha (TGF alpha, Mr 5623, pI 6.8)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service