SPR08SIA1
SmartPak® Purification Pack
For Direct-Q® 8 Water Purification Systems
Synonym(s):
SmartPak® DQ8 Purification Pack
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14
Recommended Products
Quality Level
packaging
pkg of 1 unit
manufacturer/tradename
SmartPak®
resistivity
>18.0 MΩ-cm (at 25 °C)
compatibility
for use with Direct-Q® UV
for use with Direct-Q®
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The SmartPak® DQ8 purification pack contains a combination of selective purification media to produce pure and ultrapure water directly from tap water. The SmartPak® DQ3 is an all-in-one twin cartridge removes trace impurities from tap water. The first cartridge combines pretreatment media and a reverse osmosis (RO) membrane. The second cartridge contains ion-exchange resins. This pack also contains exclusive Jetpore®, ultrapure grade mixed-bed ion-exchange resin to produce pure and ultrapure water directly from tap water. The SmartPak® needs to be periodically replaced during maintenance of the system. For use with Direct-Q® 8 Systems.
Application
This pack removes a wide variety of contaminants (organics, ions, particles...)
Features and Benefits
- Genuine consumable for the replacement of SPR08SIA1 SmartPak® DQ8 Purification Pack.
- Reliable and affordable ultrapure water solution.
- Pretreatment (activated carbon and depth filter).
- Reverse osmosis cartridge.
- Synthetic activated carbon.
- Virgin ion-exchange resins.
- Certificate of Quality available online.
Legal Information
DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
SMARTPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service