SNAP2BHMD0100
SNAP id® 2.0 Midi Blot Holders
8.5 x 13.5 cm, horizontal, flat profile for compact space storage
Synonym(s):
SNAP id® consumables, Western blot, Western blot holders
About This Item
Recommended Products
Product Name
SNAP id® 2.0 Midi Blot Holders (8.5 x 13.5 cm), The new Midi Blot Holders ensure the membrane is horizontal allowing uniform Antibody distribution. The flat profile alows for compact space allocation for storage, shipping & waste management.
Quality Level
manufacturer/tradename
SNAP id®
technique(s)
western blot: suitable
compatibility
for use with Commercially available blocking reagents
for use with Luminata Western HRP Substrates
for use with Nitrocellulose
for use with PVDF (Immobilon membranes)
for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01)
for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01)
for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001)
for use with commercially available detection reagents
detection method
chemiluminescent
colorimetric
fluorometric
shipped in
ambient
storage temp.
room temp
Related Categories
General description
Application
Legal Information
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Western blot protocol details protein transfer from gels to nitrocellulose, crucial for immunoblotting procedures in research.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service