SLHPX13
Millex™ polyethersulfone syringe filter
pore size 0.45 μm, diam. 13 mm, non-sterile, hydrophilic
Synonym(s):
0.45 μm PES syringe filter, Millex-GP syringe filter, Millipore Express® PES syringe filter, disposable syringe filter, polyethersulfone syringe filter, syringe filter
About This Item
Recommended Products
material
polyethersulfone membrane
polypropylene housing
Quality Level
Agency
suitable for DIN 38407-42
sterility
non-sterile
product line
Millex™
feature
holdup volume≤ 15 μL (after air purge)
hydrophilic
manufacturer/tradename
Millex™
parameter
10 bar max. inlet pressure (150 psi)
104 L/min flow rate
104 mL/min-cm2 water flow rate
45 °C max. temp.
technique(s)
HPLC: suitable
UV/Vis spectroscopy: suitable
analytical sample preparation: suitable
fast flow filtration: suitable
low protein-binding filtration: suitable
H
21 mm
diam.
13 mm
filtration area
0.8 cm2
volume
10 mL
color
green
pore size
0.45 μm
input
sample type aqueous solution(s)
sample type mild organic solution(s)
bubble point
≥2.0 bar, air with water at 23 °C
fitting
female Luer-Lok® inlet
male Luer outlet slip
application(s)
PFAS testing
shipped in
ambient
General description
• Non-sterile Millex syringe filters with Millipore Express PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
• Available in 0.22 µm and 0.45 µm pore sizes and two diameters to suit your application needs
• Low protein binding to minimize interaction with your sample and maximize recovery
• Polypropylene housing, offering a low level of extractables and broad chemical compatibility
Available in Multiple Pack Sizes:
SKU ending with NS (-------NS): 50 per pack
SKU ending with NL (-------NL): 100 per pack
SKU ending with NB (-------NB): 250 per pack
SKU ending with NK (-------NK): 1000 per pack
Applications:
Buffer Filtration, Clarification of Aqueous Solutions
Application
Packaging
Specifications
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Data and recommended filtration products for PFAS testing using Modified EPA 1633.
View data on fluorinated extractables, recovery, and recommended filtration products for PFAS testing using Modified EPA 537.1.
Validation of Millex® PES syringe filters in PFAS testing of water samples using EPA method 537.1.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service