SCT249
BioTracker™ Spermidine Polyamine Green Fluorescent Live Cell Probe
Human
Synonym(s):
clickable polyamine probe, live cell probe, polyamine probe, polyamine transport system probe, spermidine probe
About This Item
Recommended Products
Product Name
BioTracker™ Spermidine Polyamine Green Fluorescent Live Cell Probe,
biological source
human
Quality Level
packaging
vial of 500 μg
manufacturer/tradename
Millipore
technique(s)
cell culture | stem cell: suitable
shipped in
dry ice
storage temp.
−20°C
Application
MW: 599.44 g/mol
SPECTRAL PROPERTIES
λex: 494 nm
λem: 508 nm
Features and Benefits
Target description
Polyamine dysregulation is key in a variety of cancers. In colorectal cancer, polyamine content and biosynthesis is significantly higher when compared to normal colorectal tissue. A relationship between polyamines and their metabolites has been used to observe progression in cancers of the breast, lung, colorectal system, ovary, prostate, and pancreas.
In normal cells, there is tight regulation of polyamines through normal cellular mechanisms. One of these is the polyamine transport system (PTS). The characteristics of the PTS are not fully understood, but study of the PTS has shown that many cancer cells have high PTS activity, suggesting this system as a promising target for cancer research.
Storage and Stability
Other Notes
Legal Information
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service