SANKIT004
Sanitization Kit
For AFS® Water Purification Systems
Synonym(s):
Sanitization Kit for AFS® Water Purification Systems
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
Includes RO-Tank-Distribution loop; 4 Steps, < 10 minutes of operator time
maximum duration of 4 h
packaging
pkg of 1 kit
compatibility
for use with AFS®
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Application
Sanitization kit for use with AFS® water purification systems
Features and Benefits
Genuine consumable for the sanitization of AFS® Water Purification Systems.
Legal Information
AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service