MABF230
Anti-TIM-3, clone RMT3-23, Azide Free Antibody
clone RMT3-23, from rat
Synonym(s):
Hepatitis A virus cellular receptor 2 homolog, HAVcr-2, T-cell immunoglobulin and mucin domain-containing protein 3, TIMD-3, T-cell immunoglobulin mucin receptor 3, TIM-3, T-cell membrane protein 3
About This Item
Recommended Products
biological source
rat
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
RMT3-23, monoclonal
species reactivity
mouse
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Havcr2(171285)
General description
Immunogen
Application
Flow Cytometry Analysis: 1 µg from a representative lot detected TIM-3 in TIM-3 transfected 1X10E6 BALB/L5178Y cells.
Flow Cytometry Analysis: A representative lot detected TIM-3 in dendritic cells, macrophages, splenocytes, and hepatic mononuclear cells from acute graft-vs-host disease (aGVHD) mice (Oikawa, T., et al. (2006). J Immunol. 177(7):4281-4287.).
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected TIM-3 in liver tissue from normal and aGVHD mice (Oikawa, T., et al. (2006). J Immunol. 177(7):4281-4287.).
Immunocytochemistry Analysis: A representative lot from an independent laboratory detected TIM-3 in HMGB1-deficient MEF cells (Chiba, S., et al. (2012). Nat Immunol. 13(9):832-842.).
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected TIM-3 in 10 µg of TIM-3 transfected L5178Y cell lysate.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service