Skip to Content
Merck
All Photos(1)

Key Documents

MAB348-100UL

Anti-APP A4, a.a. 66-81 of APP (NT) antibody produced in mouse

Synonym(s):

APP A4 Detection Antibody, Anti-APP Antibody, Mouse Anti-APP A4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

Pricing and availability is not currently available.

biological source

mouse

species reactivity

fish, human, pig, monkey, mouse, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service